siRNA Details
Gene Symbol: MAGEB17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277307.1 | NP_001264236.1 | 2 | 734 | ||
XM_011545567.2 | XP_011543869.1 | 3 | 881 | |||
GenBank mRNA | DV080535.1 | - | 2 | 738 |
