siRNA Details
Gene Symbol: LOC100130698 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136612.1 | 4 | 265 | |||
GenBank mRNA | HY021529.1 | - | 4 | 265 |
Gene Symbol: LOC729815 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136625.1 | 2 | 198 |
