siRNA Details
Gene Symbol: GFY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195256.1 | NP_001182185.1 | 1 | 1161 | ||
XM_011526299.2 | XP_011524601.1 | 2 | 1273 | |||
GenBank mRNA | DN930573.1 | - | 1 | 16 |
