siRNA Details
Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104192.2 | - | 1 | 171 | ||
BC104193.2 | - | 1 | 171 | |||
BC112986.1 | - | 1 | 171 | |||
BC112987.1 | - | 1 | 171 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PTGER4P2-CDK2AP2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104192.2 | - | 1 | 171 | ||
BC104193.2 | - | 1 | 171 | |||
BC112986.1 | - | 1 | 171 | |||
BC112987.1 | - | 1 | 171 |