siRNA Details
Gene Symbol: LRRC72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195280.1 | NP_001182209.1 | 1 | 137 | ||
XM_011515057.1 | XP_011513359.1 | 1 | 190 | |||
GenBank mRNA | BF222754.1 | - | 3 | 316 |
