siRNA Details
Gene Symbol: LOC646762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024278.1 | 2 | 1873 | |||
GenBank mRNA | BC044638.1 | - | 1 | 179 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC646762 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024278.1 | 2 | 1873 | |||
GenBank mRNA | BC044638.1 | - | 1 | 179 |