siRNA Details
Gene Symbol: LOC107986805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012932.1 | XP_016868421.1 | 1 | 2108 |
Gene Symbol: LOC100132217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK128836.1 | - | 3 | 3892 |
