siRNA Details
Gene Symbol: GS1-259H13.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011515691.2 | XP_011513993.1 | 3, 4 | 322 | ||
XM_011515692.2 | XP_011513994.1 | 3, 4 | 319 | |||
XM_011515694.2 | XP_011513996.1 | 3, 4 | 310 | |||
XM_011515695.2 | XP_011513997.1 | 3, 4 | 313 | |||
XM_011515696.2 | XP_011513998.1 | 3, 4 | 260 | |||
XM_011515697.1 | XP_011513999.1 | 3, 4 | 202 | |||
XM_017011629.1 | XP_016867118.1 | 3, 4 | 245 |
