siRNA Details
Gene Symbol: LOC105376722 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022777.1 | XP_016878266.1 | 2 | 93 |
Gene Symbol: LOC100996911 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030092.1 | XP_016885581.1 | 2 | 141 | ||
XM_017030094.1 | XP_016885583.1 | 2 | 120 | |||
XM_017030095.1 | XP_016885584.1 | 2 | 120 | |||
XM_017030096.1 | XP_016885585.1 | 2 | 120 |
Gene Symbol: LOC101929235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030098.1 | XP_016885587.1 | 2 | 120 |
Gene Symbol: UBE2Q2P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003661.2 | 1 | 1132 | |||
GenBank mRNA | AK125266.1 | - | 1 | 1132 |
