siRNA Details
Gene Symbol: LINC00640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038358.1 | 2, 3 | 233 | |||
GenBank mRNA | BG198945.1 | - | 3, 4 | 340 | ||
BX111479.1 | - | 2, 3 | 85 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00640 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038358.1 | 2, 3 | 233 | |||
GenBank mRNA | BG198945.1 | - | 3, 4 | 340 | ||
BX111479.1 | - | 2, 3 | 85 |