siRNA Details
Gene Symbol: GS1-124K5.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134570.1 | 1 | 111 | |||
NR_134571.1 | 1 | 111 | ||||
NR_134572.1 | 1 | 111 | ||||
GenBank mRNA | AY726579.1 | - | 1 | 136 | ||
CT005114.1 | - | 1 | 96 | |||
HY009490.1 | - | 1 | 113 | |||
HY106786.1 | - | 1 | 81 |
