siRNA Details
Gene Symbol: SPDYE18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005250742.4 | XP_005250799.1 | 6 | 898 | ||
XM_005250743.4 | XP_005250800.1 | 5 | 753 | |||
XM_011516756.2 | XP_011515058.1 | 5 | 667 |
