siRNA Details
Gene Symbol: ZGLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103167.1 | NP_001096637.1 | 4 | 1645 | ||
GenBank mRNA | AK096830.1 | - | 4 | 1968 | ||
CK826281.1 | - | 1 | 349 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZGLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103167.1 | NP_001096637.1 | 4 | 1645 | ||
GenBank mRNA | AK096830.1 | - | 4 | 1968 | ||
CK826281.1 | - | 1 | 349 |