siRNA Details
Gene Symbol: LOC107986096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007619.1 | XP_016863108.1 | 5 | 692 | ||
GenBank mRNA | KC470081.1 | - | 4 | 632 | ||
KC470082.1 | - | 5 | 671 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC107986096 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017007619.1 | XP_016863108.1 | 5 | 692 | ||
GenBank mRNA | KC470081.1 | - | 4 | 632 | ||
KC470082.1 | - | 5 | 671 |