siRNA Details
Gene Symbol: RAD54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205263.1 | NP_001192192.1 | 1 | 285 | ||
GenBank mRNA | DA553267.1 | - | 1 | 285 |
Gene Symbol: FSBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256141.1 | NP_001243070.1 | 1 | 285 | ||
GenBank mRNA | AF007866.1 | - | 1 | 237 |
