siRNA Details
Gene Symbol: CCDC85C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144995.1 | NP_001138467.1 | 6 | 2147 | ||
XM_011536705.2 | XP_011535007.1 | 7 | 1830 | |||
GenBank mRNA | AK093969.1 | - | 1 | 290 | ||
AK123840.1 | - | 5 | 1035 | |||
BC110042.1 | - | 1 | 413 |
