siRNA Details
Gene Symbol: OOEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010890.1 | XP_016866379.1 | 1 | 54 |
Gene Symbol: OOEP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DB456122.1 | - | 1 | 128 |
