siRNA Details
Gene Symbol: FAM205C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309426.1 | NP_001296355.1 | 1, 2 | 208 | ||
NM_001309427.1 | NP_001296356.1 | 1, 2 | 208 | |||
XM_017014165.1 | XP_016869654.1 | 1, 2 | 208 | |||
GenBank mRNA | DB042441.1 | - | 1, 2 | 210 | ||
DB454411.1 | - | 1, 2 | 191 |
