siRNA Details
Gene Symbol: ZNF517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517014.2 | XP_011515316.1 | 5 | 4554 | ||
XM_011517015.2 | XP_011515317.1 | 5 | 3996 | |||
XM_011517017.2 | XP_011515319.1 | 4 | 4413 | |||
XM_017013384.1 | XP_016868873.1 | 2 | 3939 |
Gene Symbol: LOC100130027 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127225.1 | - | 5 | 2371 |
