siRNA Details
Gene Symbol: AATBC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026961.1 | 1 | 2306 | |||
GenBank mRNA | AK093410.1 | - | 1 | 100 | ||
AK128598.1 | - | 1 | 2307 | |||
BC034348.1 | - | 1 | 70 | |||
BC111558.1 | - | 1 | 2305 |
