siRNA Details
Gene Symbol: TMEM262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242631.2 | NP_001229560.1 | 1 | 180 | ||
NM_001282448.1 | NP_001269377.1 | 1 | 180 | |||
GenBank mRNA | AA416856.1 | - | 2 | 450 | ||
AI015611.1 | - | 3 | 300 | |||
AK124141.1 | - | 1 | 178 | |||
BX093670.1 | - | 1 | 135 | |||
HY032040.1 | - | 1 | 182 |
