siRNA Details
Gene Symbol: HOXB-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033202.2 | 2 | 183 | |||
NR_033203.1 | 2 | 86 | ||||
NR_033205.1 | 2 | 179 | ||||
NR_110331.1 | 2 | 183 | ||||
GenBank mRNA | BG203124.1 | - | 2 | 214 | ||
BG203609.1 | - | 2 | 209 | |||
CB046509.1 | - | 2 | 95 | |||
CF127739.1 | - | 2 | 161 |
