siRNA Details
Gene Symbol: SMIM6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162997.1 | NP_001156469.1 | 2 | 335 | ||
XM_005256903.4 | XP_005256960.1 | 3 | 1813 | |||
GenBank mRNA | AK131023.1 | - | 2 | 262 | ||
AL041162.1 | - | 1 | 524 |
