siRNA Details
Gene Symbol: LOC105370333 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132422.1 | 1 | 268 | |||
GenBank mRNA | AV692313.1 | - | 1 | 268 | ||
BG033002.1 | - | 1 | 254 | |||
CD248433.1 | - | 1 | 205 | |||
CF454479.1 | - | 1 | 215 | |||
CX784348.1 | - | 1 | 173 |
