siRNA Details
Gene Symbol: RNF212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282322.1 | NP_001269251.1 | 8 | 591 | ||
XM_011536313.2 | XP_011534615.1 | 9 | 732 | |||
XM_011536315.2 | XP_011534617.1 | 9 | 732 | |||
XM_011536316.2 | XP_011534618.1 | 9 | 732 | |||
XM_011536319.2 | XP_011534621.1 | 7 | 505 | |||
XM_011536322.2 | XP_011534624.1 | 9 | 733 | |||
XM_011536323.2 | XP_011534625.1 | 9 | 736 | |||
XM_011536328.2 | XP_011534630.1 | 7 | 508 |
