siRNA Details
Gene Symbol: RNF212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282322.1 | NP_001269251.1 | 10 | 699 | ||
XM_011536313.2 | XP_011534615.1 | 11 | 840 | |||
XM_011536314.2 | XP_011534616.1 | 9 | 764 | |||
XM_011536315.2 | XP_011534617.1 | 11 | 840 | |||
XM_011536319.2 | XP_011534621.1 | 9 | 613 | |||
XM_011536323.2 | XP_011534625.1 | 11 | 844 | |||
XM_011536324.2 | XP_011534626.1 | 9 | 765 | |||
XM_011536327.2 | XP_011534629.1 | 9 | 769 | |||
XM_011536328.2 | XP_011534630.1 | 9 | 616 | |||
XM_011536334.2 | XP_011534636.1 | 3 | 111 |
