siRNA Details
Gene Symbol: RNF212B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282322.1 | NP_001269251.1 | 12 | 796 | ||
XM_011536313.2 | XP_011534615.1 | 13 | 937 | |||
XM_011536314.2 | XP_011534616.1 | 11 | 861 | |||
XM_011536319.2 | XP_011534621.1 | 11 | 710 | |||
XM_011536328.2 | XP_011534630.1 | 11 | 713 | |||
XM_011536334.2 | XP_011534636.1 | 6 | 269 |
