siRNA Details
Gene Symbol: LINC00614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL044322.1 | - | 1 | 386 | ||
DB025290.1 | - | 1 | 451 |
Gene Symbol: ANKRD26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017015929.1 | XP_016871418.1 | 41 | 19222 |
