siRNA Details
Gene Symbol: KIAA1257 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011513050.2 | XP_011511352.2 | 9 | 1423 | ||
XM_017006939.1 | XP_016862428.1 | 10 | 1978 | |||
XM_017006940.1 | XP_016862429.1 | 10 | 1974 | |||
XM_017006941.1 | XP_016862430.1 | 10 | 1551 | |||
XM_017006942.1 | XP_016862431.1 | 10 | 1945 | |||
XM_017006943.1 | XP_016862432.1 | 10 | 1703 | |||
XM_017006944.1 | XP_016862433.1 | 9 | 1523 | |||
XM_017006945.1 | XP_016862434.1 | 9 | 1423 | |||
XM_017006946.1 | XP_016862435.1 | 8 | 1134 | |||
XM_017006947.1 | XP_016862436.1 | 7 | 1018 | |||
XM_017006948.1 | XP_016862437.1 | 9 | 1195 | |||
XM_017006949.1 | XP_016862438.1 | 8 | 1142 | |||
XM_017006950.1 | XP_016862439.1 | 8 | 6629 | |||
XM_017006951.1 | XP_016862440.1 | 8 | 6734 | |||
XM_017006952.1 | XP_016862441.1 | 7 | 912 | |||
XM_017006953.1 | XP_016862442.1 | 7 | 918 |
