siRNA Details
Gene Symbol: LOC107985527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003061.1 | XP_016858550.1 | 1 | 64 |
Gene Symbol: FLJ37786 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK095105.1 | - | 1 | 268 |
Gene Symbol: LOC105371215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510253.2 | XP_011508555.1 | 1 | 299 |
