siRNA Details
Gene Symbol: FNDC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242659.1 | NP_001229588.1 | 1 | 1124 | ||
GenBank mRNA | AB188495.1 | - | 1 | 1124 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FNDC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242659.1 | NP_001229588.1 | 1 | 1124 | ||
GenBank mRNA | AB188495.1 | - | 1 | 1124 |