siRNA Details
Gene Symbol: UST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005715.2 | NP_005706.1 | 4 | 791 | ||
XM_011535378.2 | XP_011533680.1 | 4 | 1482 | |||
XM_017010152.1 | XP_016865641.1 | 4 | 2035 | |||
GenBank mRNA | AB020316.1 | - | 4 | 598 | ||
AK292922.1 | - | 4 | 539 | |||
AK315320.1 | - | 4 | 499 | |||
BC093668.1 | - | 4 | 543 | 7939513 | ||
BC093694.1 | - | 4 | 543 | 7939539 |
