siRNA Details
Gene Symbol: LILRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278319.1 | NP_001265248.1 | 5, 6 | 1016 | ||
NM_006863.3 | NP_006854.1 | 6, 7 | 1222 | |||
NR_103501.1 | 5, 6 | 1024 | ||||
NR_103502.1 | 5, 6 | 1384 | ||||
NR_103503.1 | 5, 6 | 1140 | ||||
GenBank mRNA | AF025530.1 | - | 6, 7 | 1150 | ||
AK290919.1 | - | 6, 7 | 1166 | |||
AK310122.1 | - | 5, 6 | 743 | |||
BC093708.1 | - | 5, 6 | 996 | 7939553 | ||
BC101514.1 | - | 5, 6 | 996 | 8069020 | ||
EU915602.1 | - | 5, 6 | 985 |
