siRNA Details
Gene Symbol: LILRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278318.1 | NP_001265247.1 | 6, 7 | 962 | ||
NM_006863.3 | NP_006854.1 | 8, 9 | 1562 | |||
NR_103501.1 | 7, 8 | 1364 | ||||
GenBank mRNA | AF025529.1 | - | 6, 7 | 878 | ||
AF025530.1 | - | 8, 9 | 1490 | |||
AK290919.1 | - | 8, 9 | 1506 | |||
BC093708.1 | - | 7, 8 | 1336 | 7939553 | ||
BC101514.1 | - | 7, 8 | 1336 | 8069020 |
