siRNA Details
Gene Symbol: UPK1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281443.1 | NP_001268372.1 | 2 | 89 | ||
NM_007000.3 | NP_008931.1 | 2 | 89 | |||
XM_017026225.1 | XP_016881714.1 | 2 | 478 | |||
GenBank mRNA | AF085807.1 | - | 1 | 57 | ||
BC107097.1 | - | 1 | 62 | 40015698 | ||
BC107098.1 | - | 1 | 62 | 40015700 |
