siRNA Details
Gene Symbol: CKM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001824.4 | NP_001815.2 | 2 | 308 | ||
XM_017026240.1 | XP_016881729.1 | 1 | 193 | |||
GenBank mRNA | AK129878.1 | - | 2 | 206 | ||
AK300730.1 | - | 2 | 206 | |||
AK313281.1 | - | 2 | 206 | |||
BC007462.1 | - | 2 | 206 | 3934232 | ||
BT006793.1 | - | 1 | 133 | |||
DA896166.1 | - | 2 | 306 | |||
DC385783.1 | - | 2 | 309 |
