siRNA Details
Gene Symbol: KRBA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_213597.3 | NP_998762.1 | 1, 2 | 229 | ||
XM_005256443.2 | XP_005256500.1 | 4, 5 | 1386 | |||
XM_011523643.1 | XP_011521945.1 | 3, 4 | 1202 | |||
XM_011523644.1 | XP_011521946.1 | 2, 3 | 993 | |||
XM_017024163.1 | XP_016879652.1 | 3, 4 | 1475 | |||
GenBank mRNA | AK131219.1 | - | 1, 2 | 230 | ||
BC033390.1 | - | 1, 2 | 64 |
