siRNA Details
Gene Symbol: ZSWIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042697.1 | NP_001036162.1 | 5 | 480 | ||
NM_001042698.1 | NP_001036163.1 | 5 | 480 | |||
GenBank mRNA | AK093384.1 | - | 3 | 229 | ||
BC018807.1 | - | 5 | 451 | |||
BC044842.1 | - | 5 | 438 | |||
BC066344.1 | - | 5 | 472 | |||
BC089386.1 | - | 6 | 564 | |||
BC094855.1 | - | 7 | 762 | |||
BU075283.1 | - | 5 | 483 | |||
DQ530600.1 | - | 5 | 382 |
