siRNA Details
Gene Symbol: ZNF513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201459.1 | NP_001188388.1 | 3 | 1324 | ||
NM_144631.5 | NP_653232.3 | 4 | 1401 | |||
XM_005264142.2 | XP_005264199.1 | 4 | 1234 | |||
XM_005264143.3 | XP_005264200.1 | 3 | 788 | |||
GenBank mRNA | AB075875.1 | - | 3 | 1324 | ||
AK056765.1 | - | 2 | 706 | |||
AK096933.1 | - | 3 | 1263 | |||
AK290690.1 | - | 4 | 1375 | |||
AL833946.1 | - | 1 | 562 | |||
BC040650.1 | - | 2 | 695 | |||
BC052282.1 | - | 4 | 1383 | 5467936 |
