siRNA Details
Gene Symbol: CCDC148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301684.1 | NP_001288613.1 | 1, 2 | 347 | ||
NM_001301685.1 | NP_001288614.1 | 1, 2 | 347 | |||
NM_138803.3 | NP_620158.3 | 1, 2 | 347 | |||
XM_005246319.3 | XP_005246376.1 | 2, 3 | 234 | |||
XM_011510638.2 | XP_011508940.1 | 2, 3 | 235 | |||
XM_017003392.1 | XP_016858881.1 | 1, 2 | 347 | |||
XM_017003395.1 | XP_016858884.1 | 1, 2 | 347 | |||
XM_017003396.1 | XP_016858885.1 | 1, 2 | 347 | |||
GenBank mRNA | AF458588.1 | - | 1, 2 | 296 | ||
AK058105.1 | - | 2, 3 | 209 | |||
AK123220.1 | - | 1, 2 | 193 | |||
AK308783.1 | - | 1, 2 | 347 | |||
BC015395.1 | - | 1, 2 | 296 | |||
BC105928.1 | - | 1, 2 | 160 | |||
BC107596.1 | - | 1, 2 | 160 | 40025915 | ||
BC107597.1 | - | 1, 2 | 160 | 40025920 | ||
DA230063.1 | - | 1, 2 | 347 |
