siRNA Details
Gene Symbol: CREBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310.3 | NP_001301.1 | 2 | 443 | ||
GenBank mRNA | AB451461.1 | - | 1 | 103 | ||
AF039081.1 | - | 2 | 379 | |||
AK307202.1 | - | 3 | 319 | |||
BC104873.1 | - | 2 | 287 | 8143876 | ||
BC106052.1 | - | 2 | 384 | 6150812 | ||
BC112157.1 | - | 2 | 287 | 8327625 | ||
BY795169.2 | - | 2 | 443 | |||
CR542250.1 | - | 1 | 103 |
