siRNA Details
Gene Symbol: ZNF578 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099694.1 | NP_001093164.1 | 6 | 584 | ||
XM_017026302.1 | XP_016881791.1 | 6 | 604 | |||
GenBank mRNA | AK299106.1 | - | 6 | 584 | ||
BX955279.1 | - | 6 | 585 |
