siRNA Details
Gene Symbol: RNF187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010858.2 | NP_001010858.2 | 2 | 541 | ||
GenBank mRNA | BC012758.2 | - | 2 | 515 | ||
BC064481.1 | - | 2 | 75 | |||
BC080645.1 | - | 2 | 75 | |||
BI551408.1 | - | 2 | 546 |
