siRNA Details
Gene Symbol: CSAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102576.2 | NP_001096046.2 | 3 | 184 | ||
NM_153478.2 | NP_705611.2 | 4 | 400 | |||
GenBank mRNA | AF268418.1 | - | 3 | 154 | ||
AF268419.1 | - | 4 | 351 | |||
BC059947.1 | - | 3 | 154 | 5397842 | ||
BM810082.1 | - | 3 | 166 |
