siRNA Details
Gene Symbol: PSORS1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014068.2 | NP_054787.2 | 5 | 441 | ||
GenBank mRNA | AB031479.1 | - | 5 | 426 | ||
AF484418.1 | - | 5 | 442 | |||
AK309151.1 | - | 2 | 459 | |||
DB454051.1 | - | 3 | 228 |
