siRNA Details
Gene Symbol: METTL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113528.1 | NP_001107000.1 | 6 | 1177 | ||
NM_001297775.1 | NP_001284704.1 | 6 | 1133 | |||
NM_152636.2 | NP_689849.2 | 6 | 1177 | |||
XM_011519935.2 | XP_011518237.1 | 8 | 1398 | |||
XM_011519937.2 | XP_011518239.1 | 5 | 639 | |||
XM_011519941.2 | XP_011518243.1 | 5 | 941 | |||
XM_017017297.1 | XP_016872786.1 | 6 | 1081 | |||
XM_017017298.1 | XP_016872787.1 | 9 | 1437 | |||
XM_017017299.1 | XP_016872788.1 | 5 | 1926 | |||
XM_017017300.1 | XP_016872789.1 | 7 | 1134 | |||
XM_017017301.1 | XP_016872790.1 | 6 | 1177 | |||
XM_017017302.1 | XP_016872791.1 | 4 | 536 | |||
XM_017017303.1 | XP_016872792.1 | 8 | 1336 | |||
GenBank mRNA | AK091298.1 | - | 5 | 829 | ||
AL832668.1 | - | 5 | 830 | |||
BC030997.1 | - | 5 | 806 | 4657323 | ||
BC066900.1 | - | 6 | 1015 |
