siRNA Details
Gene Symbol: CT62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102658.1 | NP_001096128.1 | 3 | 596 | ||
XM_006720429.3 | XP_006720492.1 | 3 | 856 | |||
GenBank mRNA | AL080151.1 | - | 3 | 867 | ||
BC039359.1 | - | 3 | 592 | |||
BC048128.1 | - | 3 | 558 |
