siRNA Details
Gene Symbol: GRAMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012642.2 | NP_001012660.1 | 3, 4 | 219 | ||
XM_011521327.2 | XP_011519629.1 | 3, 4 | 328 | |||
XM_011521328.2 | XP_011519630.1 | 3, 4 | 679 | |||
XM_011521329.2 | XP_011519631.1 | 3, 4 | 196 | |||
XM_011521330.2 | XP_011519632.1 | 3, 4 | 280 | |||
XM_011521331.2 | XP_011519633.1 | 3, 4 | 219 | |||
XM_011521332.2 | XP_011519634.1 | 4, 5 | 403 | |||
XM_011521333.2 | XP_011519635.1 | 4, 5 | 420 | |||
XM_017021997.1 | XP_016877486.1 | 3, 4 | 363 | |||
XM_017021998.1 | XP_016877487.1 | 3, 4 | 587 | |||
GenBank mRNA | AK095072.1 | - | 3, 4 | 209 | ||
BC038451.1 | - | 3, 4 | 193 | |||
DA854662.1 | - | 3, 4 | 219 |
