siRNA Details
Gene Symbol: ZNF626 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076675.2 | NP_001070143.1 | 4 | 613 | ||
GenBank mRNA | AK096221.1 | - | 2 | 339 | ||
BC150651.1 | - | 4 | 524 | 9057022 | ||
BX648764.1 | - | 2 | 366 | |||
CR749233.1 | - | 4 | 565 |
