siRNA Details
Gene Symbol: C1orf168 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004303.4 | NP_001004303.3 | 5, 6 | 1478 | ||
XM_017000544.1 | XP_016856033.1 | 6, 7 | 1242 | |||
XM_017000545.1 | XP_016856034.1 | 6, 7 | 1302 | |||
GenBank mRNA | BX648439.1 | - | 5, 6 | 1154 |
